Learn More
DWK Life Sciences Plastic Hose Connection, with silicone seal (VMQ), PP complete with screw cap PBT, bent
For safe connection of hoses.
Brand:  DWK Life Sciences 286222005
4.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10293541
  • 5.70 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PP are used for lower thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/Pk.
For applications with lower thermal and chemical resistance requirements
8.6 mm