Learn More
Microspec™ Plastic Inoculation Loops
Available as aseptic manufactured or ethylene oxide sterilized.
Brand:  Microspec™ C5G
 View more versions of this product
Product Code. 16664822
  • 28.15 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Specifications
Single-use
Sterile
200 mm
Microseal Pouch
Not Autoclavable
1 μL
Blue
1 Ea.
Yes