Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
Brand: Fisherbrand™ 10 36 2401
64.77 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
- Plastic Disposable Syringe
- Polypropylene
- Luer Lock
- Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
- No silicone or oil lubricant is required in the barrel
Specifications
Polypropylene | |
10 mL | |
Luer Lock |