Learn More
Fisherbrand™ Plastic PP Syringes, Luer Slip
HPLC certified, non sterile disposable syringes made of solvent robust polypropylene
12.90 € - 32.96 €
Specifications
Material Polypropylene
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12951031
View Documents
Fisherbrand™
20 36 2406
20 mL
32.96 €
Pack of 100
12991021
View Documents
Fisherbrand™
02 36 2403
2 mL
12.90 €
Pack of 100
12911031
View Documents
Fisherbrand™
05 36 2404
5 mL
18.35 €
Pack of 100
12931031
View Documents
Fisherbrand™
10 36 2405
10 mL
23.55 €
Pack of 100
Description
Description
  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Slip
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
Specifications
Specifications
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Syringes, Luer Slip