missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Plastic Squeegee

These durably constructed squeegees are made with a polypropylene frame and a choice of either solid rubber or double foam blades for use in wash and production areas. Designed with an ACME threaded handle connection for use with most threaded plastic handles.

Brand:  Micronova™ SQRE-24

 View more versions of this product

Product Code. 13167606

  • 82.25 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. Designed for use with STAH or STAEH series plastic handles. Available in a variety of colors.

  • Durably constructed polypropylene squeegees
  • Choice of solid rubber or double foam blades
  • Excellent for use on smooth surfaces and uneven floor surfaces
  • Prevents hazardous sparks when cleaning-up flammable fluids
  • Available in 18 and 24 inch (45 and 60 cm) widths
  • Use with STAH or STAEH handles for a secure fit
  • Will not rust
Specifications

Specifications

Squeegee, ACME Threaded End
Store in cool dry area when not in use.
Reusable
STA Series Plastic Handles
1.5 in.
Red
61 x 3.8 cm (24 x 1.5 in.)
Squeegee for Floors, Walls and Windows
Foam Blade, Polypropylene Frame
3.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ Plastic Squeegee > Squeegee, Plastic Foam blade, ACME Threaded End, Red 24 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.