Learn More
KD Scientific Plastic Syringe
Economical and disposable, available in sizes ranging from 1 cc to 50 cc, Luer Slip (LS) or Luer Lock (LL) tip
Brand:  KD Scientific 780850
 View more versions of this product
Product Code. 15174194
  • 33.95 € / Pack of 25
Explore more special offers
Description
Description
  • A new line of disposable plastic sterile syringes for all scientific applications
  • Norm-ject syringes are the ideal solution for any situation
  • Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
Specifications
Polypropylene (Barrel)
Yes
Single-use
KD Scientific Syringe Pumps
Yes
No Needle Included
Syringe
1 mL
Tuberculin Luer Slip
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KD Scientific Plastic Syringe > 1 cc, tuberculin