missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Plastic TC Clamp

For use with the ÄKTA™ Pilot chromatography system. Cytiva Plastic TC Clamp is a white 25mm tri-clamp for providing a sanitary, secure connection.

Brand:  Cytiva 18-1169-18

Product Code. 11380062

  • 321.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • White 25mm tri-clamp
  • For use with the ÄKTA™ Pilot chromatography system
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
1 Pack
Plastic
25 mm connection
ÄKTA Pilot Chromatography System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers