Learn More
Buerkle™ LaboPlast™ Plastic Splinter-Proof Titrating Burettes
Allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet. Buerkle™ Titrating Burets with Shatter Protection has a plastic sheathing which increases resistance to breaki
Brand:  Buerkle™ 9695-3025
83.83 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11981501
  • 97.70 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Burette with splinter-proof sheathing
  • Adjusted to Ex
  • Burette made of borosilicate glass 3.3
  • Shatter-proof, resistant to chemicals
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with medium: PE, PP, natural rubber, borosilicate glass
  • Fast acting burette with Schellbach stripes based on Dr. Schilling's design
  • Plastic sheathing which increases resistance to breaking and protects against splinters
  • Allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet
Specifications
Specifications
25 mL
65 cm
0.05 mm
DIN
Plastic-coated Borosilicate Glass
Storage bottle and Immersion tube, splitter-proof sheathing, Schellbach-stripes
Titrating Buret