missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Plastic Cylinder and Equipment Brushes

These lightweight, durable equipment brushes are made from FDA approved, BPA-free base resins with polyester bristles and attach to plastic threaded handles. Available dome-shaped (Cylinder Brush) and oblong shaped (Kettle Brush) for specialized cleaning.

Brand:  Micronova™ BRWH-02

 View more versions of this product

Product Code. 12725745

  • 77.45 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket

Requires : STAH or STAEH series handles
This item is not returnable. View return policy

Description

Description

The Micronova Plastic Cylinder and Equipment Brushes are available in two varieties with optional brush covers for fine cleaning and sanitization. The Cylinder Brush is dome-shaped for small tanks, carboys, duct work, and other cylinders, optional use with BRSC6-02 brush cover. The Kettle Brush is oblong shaped for small tanks, carboys and other production equipment, optional use with BRSC6-41 brush cover.

  • Lightweight, durable equipment brushes with polyester bristles
  • Ideal for small tanks, carboys, duct work, and other cylinders
  • Made from FDA approved, BPA-free base resins
  • Cylinder Brush is a round/ dome-shaped brush
  • Kettle Brush is an oblong shaped brush
  • Brush covers available for fine cleaning and sanitization
  • FDA approved for indirect food contact
  • ACME thread for attachment to STA series handles
  • Optional fabric covers available improve fine cleaning and sanitizing
Specifications

Specifications

Cylinder Brush with Acme Thread
Plastic
Non-sterile
Keep at ambient temperature
White
7 in.
17.8 cm
Used to Clean The Bottoms and Sides of Rounded Vessels
7 in.
1 Ea.
Polyester
Acme Thread
Not Autoclavable
Certificate of Conformance upon request
Round Cylinder Brush
7 in.
Reusable
STAH and STAEH series handles
17.8 cm
Chemicals
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.