missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Finned Plug for Disposable Tubes

Product Code. p-9490008
Click to view available options
Color:
Neutral
Red
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11371195

Brand: Kartell™ 8830900

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed with flexible fins to create a snug fit.

  • Made from polyethylene
  • Available in different colors

Specifications

Certifications/Compliance CE-IVD
Product Type Finned Plug
Material Polyethylene
Color Neutral
For Use With (Equipment) 11 to 13 mm Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.