Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
For use with plastic or rubber tubing.
Brand:  Kartell™ 0141600
 View more versions of this product
Product Code. 15598265
  • 229.00 € / Pack of 100
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sturdy shape provides strength
  • Chemical and thermal resistant
Specifications
Specifications
CE IVD
Autoclavable
100/Pk.
4-way cross
15 mm
Polypropylene
11.6mm Bore
Natural