missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polypropylene Desiccator Plates

Useful as support for crucibles, petri dishes, etc.

Brand:  Kartell™ 0055100

Product Code. 11929888

  • 77.45 € / Pack of 10
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from polypropylene
  • Used at room temperature
Specifications

Specifications

7 mm
Desiccator Plate
Polypropylene
10/Pk.
Depth: 140.5mm
Kartell Filtration and Vacuum Desiccator, Part Number: 10365821
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.