Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
Designed for use with hose system and vacuum lines.
Brand:  Kartell™ 0052600
 View more versions of this product
Product Code. 12543307
  • 190.00 € / Pack of 100
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
14 mm
Polypropylene
100/Pk.
Y
Untapered Y Tubing Connector
Autoclavable
Useful for any hoses system and vacuum line
9.7mm bore