Learn More
BD PlastiPak™ Syringe with Luer Lock
Used for injections and sampling.
Brand:  BD 305959
Product Code. 15544835
  • 63.34 € / Pack of 100
Edge
Explore more special offers
Description
Description
  • 10mL capacity with 0.2mL graduations
  • Sterile and disposable
  • Polypropylene construct
  • Clear barrel for perfect visibility of syringe contents
  • Special silicone lubrication ensures that plunger moves smoothly and evenly
  • Sterilized by ethylene oxide
Certifications
EN ISO 7886-1
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.2 mL
10 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD PlastiPak™ Syringe with Luer Lock > 10mL; 100/pk.