Learn More
Thermo Scientific™ Plate Count Agar
Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.
17.65 €
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
11973612
|
Thermo Scientific™
PO0158A |
10/Pk. | 90mm Monoplate | Plate Count Agar | United Kingdom |
17.65 €
Pack of 10 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
Description
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
This standard medium meets the formulation of APHA, AOAC and PHLS.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Plate Count Agar | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 14 Weeks