missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces

Product Code. 10788944
Click to view available options
Color:
Black
Clear
White
Product Type:
Breakable Module F8 (x12) in Frame
Lock-Well Module C8 (x12) in Frame
Microplate
Solid Module F8 (x12) in Frame
Surface Treatment:
Glutathione
Streptavidin
Total Volume (Metric) Well:
120 μL
350 μL
400 μL
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 10788944

Brand: Thermo Scientific™ 436020

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.

Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.

Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)

  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 15 Cases
Binding Type Affinity
Color Clear
Product Type Solid Module F8 (x12) in Frame
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment Streptavidin
Total Volume (Metric) Well 400 μL
Well Shape Round, Flat Bottom (F-well)
No. per Case 15
No. per Pack 1
Plate Blocking Unspecified
Quantity 15/Cs.
Target Molecule Biotinylated
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.