Learn More
Grant Instruments™ Platform For V-32 Vortex Mixer
Creates versatility with platforms that accommodate a variety of tube sizes.
Brand:  Grant Instruments™ PV6-10
 View more versions of this product
Product Code. 15251226
  • 50.40 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with V-32 Vortex Mixer
Replacement Platform PV-32:
  • Fits 3 tube sizes:
    • 16 x 1.5mL
    • 8 x 0.5mL
    • 8 x 0.2mL
Universal 6-socket platform PV-6:
  • 6 x 10mL tubes
  • 15mm maximum tube dia.
Specifications
Specifications
V-32
6