Learn More
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators
Includes clamps to keep flasks and beakers from spilling. Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators are interchangeable to accommodate a variety of vessels.
449.00 € - 754.00 €
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11431068
|
Grant Instruments™
P16-250 |
16 x 250 to 300 mL Flasks |
692.00 €
Each |
In Stock
Log in to see stock. |
|||||
11421068
|
Grant Instruments™
P30-100 |
30 x 100 to 150 mL Flasks |
754.00 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
11451068
|
Grant Instruments™
P6-1000 |
6 x 1000 mL Flasks |
605.00 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
11441068
|
Grant Instruments™
P9-500 |
9 x 500 mL Flasks |
449.00 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
- Includes clamps to keep vessels secure
- For use with
- ES-80 Shaker Incubator
- PSU 20i Orbital Shaker
Specifications
PSU-20i Orbital Platform Shaker |