Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
Brand:  Grant Instruments™ PRS-14
 View more versions of this product
Product Code. 11421028
  • 145.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
  • Hold as few as 8 test tubes or as many as 48.
  • Accommodates a variety of capacities
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14 tubes up to 30mm, tube volumes up to 50mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators > For Use With: 14 tubes up to 30mm, tube volumes up to 50mL