missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Platform Rocker Stacking Bars
Accessory for use with Digital Tube Rocker, 120V and Digital Tube Rocker, 230V
Brand: Thermo Scientific™ 88882161
This item is not returnable.
View return policy
Description
Accessory for use with Digital Tube Rocker, 120V and Digital Tube Rocker, 230V
Specifications
Stainless Steel | |
Double Layer Tray (88882148), Basic Vortex Mixer, 230V (88882012), Rubber Strips (88882103) | |
4 stacking bars for 2nd platform rocker tray installation |
12 x 108 mm | |
Digital Platform Rocker 120V (88882013), Digital Platform Rocker 230V (88882014) |