missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Platform trolley, steel, foldable, 500kg capacity
Brand: Manutan A340306
This item is not returnable.
View return policy
Description
Highly durable welded steel trolleys with a foldable handle. Non-slip PVC platform surface, ensures maximum grip of load when in transit. All round buffer strip protects walls and door frames when the truck is in use. Welded steel construction.- Dimensions (W x L): 790mm x 1240mm
- Handle height: 1040mm
- Load capacity: 500kg
- Weight: 44.5kg
- Foot operated handle release bar
- Two fixed and two rear swivel non-marking tyres.