missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 10% on carbon, dry, Thermo Scientific Chemicals

45.53 € - 838.17 €

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11320064
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L00469.03
1 g
45.53 €
1g
Please to purchase this item. Need a web account? Register with us today!
11330064
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L00469.06
5 g
154.77 €
5g
Please to purchase this item. Need a web account? Register with us today!
11340064
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L00469.14
25 g
838.17 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

Solubility
Insoluble in water.

Notes
Store in a cool location. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

7440-06-4
MFCD00011179
14,7529
Insoluble in water.
[Pt]
195.08
CHEBI:33400
Platinum, 10% on carbon, dry
Pt
UN1325
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'rtecsNumber' : TP2160000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only