missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum 20 wt% Rhodium wire, 0.25mm (0.01in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
160.80 € - 624.11 €
Specifications
Chemical Name or Material | Platinum 20 wt% Rhodium wire |
---|---|
Name Note | 0.25mm (0.01 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh; 80:20 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00198186 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349388
|
Thermo Scientific Alfa Aesar
043274.BU |
25 cm |
160.80 €
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359388
|
Thermo Scientific Alfa Aesar
043274.BY |
100 cm |
624.11 €
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 20 wt% Rhodium wire | |
Pt:Rh; 80:20 wt% | |
MFCD00198186 | |
Odorless | |
Wire |
0.25mm (0.01 in.) dia. | |
(metals basis) | |
99.9% | |
≈0.24g/25cm |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only