Learn More
Platinum, 3% on carbon paste, B103032-3, Thermo Scientific™
24.38 € - 232.72 €
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
7440-06-4 | |
Powder | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
Odorless |
Black | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Platinum, 3% on carbon paste, B103032-3 |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'einecsNumber' : 231-116-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.