Learn More
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Material For Gas Sensors
87.70 € - 325.00 €
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492257
|
Thermo Scientific Alfa Aesar
045445.06 |
5 g |
87.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15466925
|
Thermo Scientific Alfa Aesar
045445.14 |
25 g |
325.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum, 5% on activated carbon paste is used for a broad range of reactions common to palladium on carbon catalysts. it is utilized for hydrogenolysis reactions using molecular hydrogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
7440-06-4 | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
Powder |
Powder | |
MFCD00011179 | |
Insoluble in water. | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Platinum, 5% on graphite |
Safety and Handling
H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-116-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only