missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on carbon, dry, Thermo Scientific Chemicals

36.88 € - 1591.92 €

Identifiants chimiques

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
Consulter d'autres spécifications

Produits 4
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
11339216
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11186.03
1 g
36.88 €
1g
Please to purchase this item. Need a web account? Register with us today!
11429133
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11186.06
5 g
118.59 €
5g
Please to purchase this item. Need a web account? Register with us today!
11439133
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11186.14
25 g
464.31 €
25g
Please to purchase this item. Need a web account? Register with us today!
11329216
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11186.22
100 g
1591.92 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts). Insoluble in water, mineral acids, and organic acids.

Notes
Do not store together with acids. Incompatible with Strong acids, strong oxidizing agents & organics Solvent vapors. Store under inert gas.
Spécification

Identifiants chimiques

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Spécification

7440-06-4
MFCD00011179
14,7529
Soluble in boiling aqua regia (reacts),and molten alkali cyanides (reacts). Insoluble in water,mineral acids,and organic acids.
[Pt]
195.08
CHEBI:33400
Pt
UN1325
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Platinum, 5% on carbon, Dry
Vidéos
Sécurité et traitement

Sécurité et traitement

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'rtecsNumber' : TP2160000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Certificats

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis