missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on carbon paste, B501018-5, Thermo Scientific™

81.20 € - 678.00 €

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427647
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H36310.06
5 g
81.20 €
5g
Please to purchase this item. Need a web account? Register with us today!
15437647
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H36310.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15447647
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H36310.22
100 g
678.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

7440-06-4
Solid
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Odorless
Gray
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Platinum, 5% on carbon paste, B501018-5
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications