Learn More
Platinum black, 98% Pt, Thermo Scientific Chemicals
351.75 € - 1479.36 €
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10410861
|
Thermo Scientific Acros
195210010 |
1 g | Glass bottle |
351.75 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10062470
|
Thermo Scientific Acros
195210050 |
5 g | Glass bottle |
1479.36 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
White | |
1772.0°C | |
21.45 | |
Pt | |
01,890; 02,332 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
≥97.5% (Pt) | |
Platinum black, 98% Pt |
7440-06-4 | |
3827.0°C | |
Pt | |
MFCD00011179 | |
15, 7644 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
195.08 | |
Powder |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 231-116-1
missing translation for 'rtecsNumber' : TP2160000
missing translation for 'tsca' : TSCA
- Platinum black
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.