missing translation for 'onlineSavingsMsg'
Learn More

Platinum black, Thermo Scientific Chemicals

Product Code. 11336438
Click to view available options
Quantity:
0.1 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: S.A. 10m2/g

Platinum black is used in polymer electrolyte fuel cells, microbial fuel cells, and phosphoric acid fuel cell applications. It is also used as a recombination catalyst for hydrogen elimination. It is also used as a building block for sensor elements. It is used as a catalyst and as a gas absorbent. It is also useful for the electrochemical detection of hydrogen peroxide and nitrite in microchannel. It is useful for the anodic oxidation of ammonia in aqueous potassium hydroxide electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Gray
Appearance Dark gray
Quantity 5 g
Solubility Information Soluble in aqua regia. Insoluble in water,mineral and organic acids.
Odor Odorless
Form Powder
Chemical Name or Material Platinum black

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'