missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Crucible
Available in 15mL and 50mL capacities
Brand: N/A 23IR
This item is not returnable.
View return policy
Description
Quoted masses are approximate.Approximate process temperature 1,200°C and maximum temperature is 1,400°C (for short time of 10min)
Specifications
Platinum | |
1Each | |
28mm | |
Crucible |
1200°C | |
15 mL | |
32mm |