missing translation for 'onlineSavingsMsg'
Learn More
Platinum foil, 0.025mm (0.001in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11366429

Platinum foil, 0.025mm (0.001in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11366429
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy

Product Code. 11366429

Brand: Thermo Scientific Alfa Aesar 011510.FI

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 1.34g/25x25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color White
Appearance Silvery white foil
Density 21.45 g/cm3 at 20°C
Assay Percent Range 99.99% (Metals basis)
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Chemical Name or Material Platinum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum foil, 0.025mm (0.001in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.