missing translation for 'onlineSavingsMsg'
Learn More

Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11391358
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color White
Appearance Silvery white guage
Density 21.45g/cm3 at 20°C
Assay Percent Range 99.9%
Quantity 25 x 25 mm
Solubility Information Insoluble in water. Dissolves in aqua regia
Mesh Size 100 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Platinum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'