Learn More
Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals
71.25 € - 2328.59 €
Chemical Identifiers
CAS | 10025-65-7 |
---|---|
Molecular Formula | Cl2Pt |
Molecular Weight (g/mol) | 265.98 |
MDL Number | MFCD00011181 |
InChI Key | CLSUSRZJUQMOHH-UHFFFAOYSA-L |
Synonym | Platinous chloride |
IUPAC Name | platinum(2+) dichloride |
SMILES | [Cl-].[Cl-].[Pt++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10267593
|
Thermo Scientific Acros
195382500 |
250 mg | Glass bottle |
71.25 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10318050
|
Thermo Scientific Acros
195380010 |
1 g | Glass bottle |
170.85 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042590
|
Thermo Scientific Acros
195380050 |
5 g | Glass bottle |
521.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954191
|
Thermo Scientific Acros
195380250 |
25 g | Glass Bottle |
2328.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-65-7 | |
265.98 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride |
Cl2Pt | |
MFCD00011181 | |
Platinous chloride | |
[Cl-].[Cl-].[Pt++] |
Specifications
581.0°C | |
Brown to Gray-Green | |
Cl2Pt | |
MFCD00011181 | |
15,7643 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid,insoluble in alcohol and ether | |
[Cl-].[Cl-].[Pt++] | |
265.98 | |
73% (Pt) | |
6.0500g/mL |
10025-65-7 | |
Powder | |
PtCl2 | |
02,272 | |
Platinous chloride | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride | |
266 | |
6.05 | |
Platinum(II) chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-034-1
- Platinum(II) chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.