Learn More
Platinum(II) cyanide, 98+%, Thermo Scientific™
141.00 € - 618.00 €
Chemical Identifiers
CAS | 592-06-3 |
---|---|
Molecular Formula | C2N2Pt |
MDL Number | MFCD00016288 |
Synonym | platinum ii cyanide, acmc-1axcr, platinumdicarbonitrile, platinum cyanide pt cn 2 7ci,8ci,9ci, platinum ii cyanide trace metals basis |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
592-06-3 | |
C2N2Pt | |
MFCD00016288 | |
platinum ii cyanide, acmc-1axcr, platinumdicarbonitrile, platinum cyanide pt cn 2 7ci,8ci,9ci, platinum ii cyanide trace metals basis |
Specifications
592-06-3 | |
Pt(CN)2 | |
UN1588 | |
247.13 | |
Platinum(II) cyanide |
C2N2Pt | |
MFCD00016288 | |
platinum ii cyanide, acmc-1axcr, platinumdicarbonitrile, platinum cyanide pt cn 2 7ci,8ci,9ci, platinum ii cyanide trace metals basis | |
≥98% |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 209-743-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures