missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min, Thermo Scientific Chemicals

Product Code. 11376548
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Platinum(II) iodide is used in pharmaceuticals and medical applications. It is also used to form cis and trans complexes with propan-1-amine. It is utilized in the catalytic hydration reaction of alkynes such as 2-hexyne, 4-methyl-2-pentyne, and 4,4dimethyl-2- pentyne . It is also used in the preparation of diiodo(2,9-dimethyl-1,10-phenanthroline)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-39-8
Molecular Formula I2Pt
Molecular Weight (g/mol) 448.89
MDL Number MFCD00011183
InChI Key ZXDJCKVQKCNWEI-UHFFFAOYSA-L
Synonym platinum ii iodide, platinum iodide pti2, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin
IUPAC Name platinum(2+) diiodide
SMILES [I-].[I-].[Pt++]

Specifications

Melting Point 325°C (decomposition)
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water,alkali iodides.
Formula Weight 448.9
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pt 43.0% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Platinum(II) iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'