missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

189.95 € - 703.50 €

Specifications

Chemical Name or Material Platinum Iridium foil
Name Note 0.025mm (0.001 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 80:20 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD02091683
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394757
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041805.FF
25 x 25 mm
189.95 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11305939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041805.FI
50 x 50 mm
703.50 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Iridium foil
Pt:Ir; 80:20 wt%
MFCD02091683
Odorless
Foil
0.025mm (0.001 in.) thick
(metals basis)
99.9%
≈0.34g/25x25mm
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only