missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.

305.52 € - 600.99 €

Specifications

Chemical Name or Material Platinum Iridium foil
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683
Solubility Information Insoluble in water.
Percent Purity 99.9%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354757
View Documents
Thermo Scientific Alfa Aesar
041802.FF
25 x 25 mm
305.52 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11375929
View Documents
Thermo Scientific Alfa Aesar
041802.FI
50 x 50 mm
600.99 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Iridium foil
MFCD02091683
99.9%
Pt:Ir; 90:10 wt%
Insoluble in water.
Tube
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only