missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 041803.FF

272.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11364757

  • 273.36 € / 25mm

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Platinum Iridium foil
Pt:Ir; 90:10 wt%
MFCD02091683
Insoluble in water.
Odorless
Foil
0.05mm (0.002 in.) thick
(metals basis excluding precious metals)
25 x 25 mm
99.9%
≈0.67g/25x25mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only