missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

647.22 € - 1562.78 €

Specifications

Chemical Name or Material Platinum Iridium foil
Molecular Formula Pt:Ir; 80:20 wt%
MDL Number MFCD02091683
Percent Purity 99.9%
Packaging Tube
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314767
View Documents
Thermo Scientific Alfa Aesar
041807.FF
25 x 25 mm
647.22 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11325939
View Documents
Thermo Scientific Alfa Aesar
041807.FI
50 x 50 mm
1562.78 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Iridium foil
MFCD02091683
Tube
Pt:Ir; 80:20 wt%
99.9%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only