missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
365.82 € - 1159.77 €
Specifications
Chemical Name or Material | Platinum Iridium foil |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD02091683 |
Percent Purity | 99.9% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384757
|
Thermo Scientific Alfa Aesar
041804.FF |
25 x 25 mm |
365.82 €
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395929
|
Thermo Scientific Alfa Aesar
041804.FI |
50 x 50 mm |
1159.77 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium foil | |
(metals basis) | |
99.9% | |
Foil |
Pt:Ir; 90:10 wt% | |
MFCD02091683 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only