missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire, Thermo Scientific Chemicals

Filtration, electrodes

Brand:  Thermo Scientific Alfa Aesar 040934.FF

 View more versions of this product

Product Code. 11375237

  • 276.38 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Platinum Iridium gauze
Pt:Ir; 90:10 wt%
25 x 25 mm
Odorless
Woven from 0.043mm (0.0017 in.) dia. wire
MFCD02091683
150 Mesh
Wire Cloth
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only