missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium wire, 0.152mm (0.006in) dia, hard, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
65.83 € - 240.20 €
Specifications
Chemical Name or Material | Platinum Iridium wire |
---|---|
Name Note | 0.152mm (0.006 in.) dia., Hard |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 80:20 wt% |
Assay Percent Notes | (metals basis excluding precious metals) |
MDL Number | MFCD02091683 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349498
|
Thermo Scientific Alfa Aesar
043331.BS |
10 cm |
65.83 €
10cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339498
|
Thermo Scientific Alfa Aesar
043331.BW |
50 cm |
240.20 €
50cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 80:20 wt% | |
MFCD02091683 | |
Odorless | |
Wire |
0.152mm (0.006 in.) dia., Hard | |
(metals basis excluding precious metals) | |
99.9% | |
≈0.04g/10cm |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only