missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium wire, 1.0mm (0.04in) dia, hard, Thermo Scientific Chemicals
603.00 € - 4708.43 €
Specifications
Chemical Name or Material | Platinum Iridium wire |
---|---|
Name Note | 1.0mm (0.04 in.) dia., Hard |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 80:20 wt% |
MDL Number | MFCD02091683 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489960
|
Thermo Scientific Alfa Aesar
044732.BQ |
5 cm |
603.00 €
5cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499960
|
Thermo Scientific Alfa Aesar
044732.BS |
10 cm |
1165.80 €
10cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409970
|
Thermo Scientific Alfa Aesar
044732.BW |
50 cm |
4708.43 €
50cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum Iridium wire, 1.0mm (0.04in) dia, hard is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 80:20 wt% | |
Insoluble in water. | |
≈0.85g/5cm |
1.0mm (0.04 in.) dia., Hard | |
MFCD02091683 | |
Odorless | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only