missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals

181.91 € - 687.42 €

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331688
SDS View Documents
Thermo Scientific Alfa Aesar
011045.03
1 g
181.91 €
1g
Please to purchase this item. Need a web account? Register with us today!
11321688
SDS View Documents
Thermo Scientific Alfa Aesar
011045.06
5 g
687.42 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13454-96-1
336.88
FBEIPJNQGITEBL-UHFFFAOYSA-J
26031
Cl[Pt](Cl)(Cl)Cl
Cl4Pt
MFCD00011182
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
tetrachloroplatinum

Specifications

370°C (decomposition)
Red to Brown
99.9% (metals basis)
MFCD00011182
Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether.
Cl[Pt](Cl)(Cl)Cl
336.88
336.9
4.303
13454-96-1
Crystalline
Cl4Pt
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
FBEIPJNQGITEBL-UHFFFAOYSA-J
tetrachloroplatinum
26031
Hygroscopic
Platinum(IV) chloride, Pt 57% min.
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H314-H317-H334

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 236-645-1

missing translation for 'rtecsNumber' : TP2275500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.