missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™

38.68 € - 1028.12 €

Chemical Identifiers

CAS 7790-46-7
Molecular Formula I4Pt
MDL Number MFCD00016292
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313998
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040401.01
0.25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11323998
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040401.06
5 g
1028.12 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-46-7
I4Pt
MFCD00016292

Specifications

7790-46-7
Powder
I4Pt
702.7
Platinum(IV) iodide
Black
99.95% (metals basis)
MFCD00016292
Black powder
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III

missing translation for 'einecsNumber' : 232-207-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only