missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals

83.11 € - 985.91 €

Chemical Identifiers

CAS 1314-15-4
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00011184
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343998
View Documents
Thermo Scientific Alfa Aesar
040402.01
0.25 g
83.11 €
0.25g
Please to purchase this item. Need a web account? Register with us today!
11363998
View Documents
Thermo Scientific Alfa Aesar
040402.03
1 g
249.24 €
1g
Please to purchase this item. Need a web account? Register with us today!
11353998
View Documents
Thermo Scientific Alfa Aesar
040402.06
5 g
985.91 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum(IV) oxide is used as catalyst for hydrogenation reactions. It is applied in the hydrogenation of alkenes to alkanes, nitro compounds to amines and ketones to alcohols. It is also involved in the hydrogenolysis of cyclopropyl group in to an isopropyl group and in selective oxidation of primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-15-4
227.08
YKIOKAURTKXMSB-UHFFFAOYSA-N
O=[Pt]=O
O2Pt
MFCD00011184
dioxoplatinum

Specifications

450°C
Brown
O2Pt
MFCD00011184
14,7527
YKIOKAURTKXMSB-UHFFFAOYSA-N
dioxoplatinum
227.09
Premion™
Odorless
(metals basis)
1314-15-4
Powder
PtO2
UN1479
Soluble in caustic potash solution. Insoluble in water,acid,aquaregia.
O=[Pt]=O
227.08
99.95%
Pt 84.4% min.
10.2 g/mL
Platinum(IV) oxide, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire
oxidizer.

P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c

H271

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: I

missing translation for 'einecsNumber' : 215-223-0

missing translation for 'rtecsNumber' : TP2506020

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.