missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%, Thermo Scientific Chemicals

Product Code. 11311698
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Platinum(IV) oxide hydrate is used in the conversion of ketones to methylenes by hydrogenolysis of enol triflates under neutral conditions. It is used as a catalyst for hydrosilylation of olefins with various alkyl dialkoxy silanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52785-06-5
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00066964
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O

Specifications

Color Brown
Physical Form Powder
Quantity 0.25 g
Linear Formula PtO2·xH2O
UN Number UN1479
Merck Index 14,7527
Formula Weight 227.09 (Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 71 to 75%
Assay (metals basis)
Chemical Name or Material Platinum(IV) oxide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'