Learn More
Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%, Thermo Scientific Chemicals
62.60 € - 599.00 €
Chemical Identifiers
CAS | 52785-06-5 |
---|---|
Molecular Formula | O2Pt |
Molecular Weight (g/mol) | 227.08 |
MDL Number | MFCD00066964 |
InChI Key | YKIOKAURTKXMSB-UHFFFAOYSA-N |
IUPAC Name | dioxoplatinum |
SMILES | O=[Pt]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311698
|
Thermo Scientific Alfa Aesar
011049.01 |
0.25 g |
62.60 €
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331698
|
Thermo Scientific Alfa Aesar
011049.03 |
1 g |
179.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321698
|
Thermo Scientific Alfa Aesar
011049.06 |
5 g |
599.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum(IV) oxide hydrate is used in the conversion of ketones to methylenes by hydrogenolysis of enol triflates under neutral conditions. It is used as a catalyst for hydrosilylation of olefins with various alkyl dialkoxy silanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52785-06-5 | |
227.08 | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
O=[Pt]=O |
O2Pt | |
MFCD00066964 | |
dioxoplatinum |
Specifications
52785-06-5 | |
Powder | |
PtO2·xH2O | |
UN1479 | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
dioxoplatinum | |
227.09 (Anhydrous) | |
Pt 71 to 75% | |
Platinum(IV) oxide hydrate |
Brown | |
O2Pt | |
MFCD00066964 | |
14,7527 | |
O=[Pt]=O | |
227.08 | |
99.9% | |
(metals basis) |
Safety and Handling
GHS H Statement
H272
May intensify fire
oxidizer.
GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
missing translation for 'rtecsNumber' : TP2506020
missing translation for 'tsca' : TSCA
RUO – Research Use Only