Learn More
Platinum(IV) oxide hydrate, typical Pt-content 79-84%, Thermo Scientific Chemicals
157.00 € - 2533.00 €
Chemical Identifiers
CAS | 52785-06-5 |
---|---|
Molecular Formula | O2Pt |
Molecular Weight (g/mol) | 227.08 |
MDL Number | MFCD00066964 |
InChI Key | YKIOKAURTKXMSB-UHFFFAOYSA-N |
Synonym | Adams' catalyst |
IUPAC Name | dioxoplatinum |
SMILES | O=[Pt]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10552231
|
Thermo Scientific Acros
211920010 |
1 g | Glass bottle |
157.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753211
|
Thermo Scientific Acros
211920050 |
5 g | Glass bottle |
577.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10629103
|
Thermo Scientific Acros
211920250 |
25 g | Glass bottle |
2533.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52785-06-5 | |
227.08 | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
dioxoplatinum |
O2Pt | |
MFCD00066964 | |
Adams' catalyst | |
O=[Pt]=O |
Specifications
450.0°C | |
Brown to Black | |
O2Pt | |
MFCD00066964 | |
01,890; 15,268 | |
Adams' catalyst | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
dioxoplatinum | |
227.09 | |
Platinum(IV) oxide hydrate |
52785-06-5 | |
Powder | |
PtO2 | |
1479 | |
14, 7527 | |
Solubility in water: insoluble. Other solubilities: soluble in conc. acids and dil.sol. of koh | |
O=[Pt]=O | |
227.08 | |
79 to 84% Pt |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
- Platinum(IV) oxide hydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only