Learn More
Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis), Thermo Scientific Chemicals
54.10 € - 824.00 €
Chemical Identifiers
CAS | 12137-21-2 |
---|---|
Molecular Formula | H2O3Pt |
Molecular Weight (g/mol) | 245.10 |
MDL Number | MFCD00066964 |
InChI Key | SPGAMGILENUIOF-UHFFFAOYSA-N |
Synonym | dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o |
PubChem CID | 11139377 |
SMILES | O.O=[Pt]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773211
|
Thermo Scientific Acros
211932500 |
250 mg | Glass bottle |
54.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276600
|
Thermo Scientific Acros
211930010 |
1 g | Glass bottle |
181.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249603
|
Thermo Scientific Acros
211930050 |
5 g | Glass bottle |
824.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
12137-21-2 | |
245.10 | |
SPGAMGILENUIOF-UHFFFAOYSA-N | |
11139377 |
H2O3Pt | |
MFCD00066964 | |
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o | |
O.O=[Pt]=O |
Specifications
12137-21-2 | |
Powder | |
H2O3Pt | |
MFCD00066964 | |
10, 7398 | |
Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh | |
O.O=[Pt]=O | |
245.10 | |
245.1 | |
Trace Metal Basis |
Brown to Black | |
99.8% | |
PtO2·H2O | |
01,890 | |
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o | |
SPGAMGILENUIOF-UHFFFAOYSA-N | |
dioxoplatinum;hydrate | |
11139377 | |
99.80% | |
Platinum(IV) oxide monohydrate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
- Platinum(IV) oxide monohydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only